Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOPP3
Entrez Gene ID
842708
Gene Name
type one serine/threonine protein phosphatase 3
Gene Aliases
AT1G64040, type one serine/threonine protein phosphatase 3
Chromosome Location
Chr.1: 23758241 - 23760663 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105077.4 NP_176587.1 1 - 2 567 88
GenBank mRNA AY081338.1 - 1 - 2 403 88
AY128830.1 - 1 - 2 178 88
M93410.1 - 1 - 2 213 88

View Full Product Details