Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC5
Entrez Gene ID
827001
Gene Name
plastid transcriptionally active 5
Gene Aliases
AT4G13670, F18A5.60, F18A5_60, plastid transcriptionally active 5
Chromosome Location
Chr.4: 7948579 - 7951022 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340872.1 NP_001328390.1 5 - 6 1396 66
NM_117441.4 NP_193103.2 5 - 6 1128 66
GenBank mRNA BT029541.1 - 5 - 6 1063 66

View Full Product Details