Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDS3
- Entrez Gene ID
-
827061
- Gene Name
- phytoene desaturase 3
- Gene Aliases
-
AT4G14210, DL3145C, FCAALL.28, PDE226, PDS, PHYTOENE DESATURASE, PIGMENT DEFECTIVE 226, phytoene desaturase 3
- Chromosome Location
-
Chr.4: 8190190 - 8195320 on Build TAIR10.1
- Gene Symbol
- ACT2
- Entrez Gene ID
-
821411
- Gene Name
- actin 2
- Gene Aliases
-
AT3G18780, DEFORMED ROOT HAIRS 1, DER1, ENHANCER OF LRX1 2, ENL2, FIZ2, FRIZZY AND KINKED SHOOTS 2, LIGHT STRESS-REGULATED 2, LSR2, actin 2
- Chromosome Location
-
Chr.3: 6474710 - 6477204 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230311.1 |
- |
- |
161 |
71 |
View Full Product Details