Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAA3
Entrez Gene ID
827233
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT4G15570, DL3825W, FCAALL.334, MAGATAMA 3
Chromosome Location
Chr.4: 8892607 - 8899061 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341063.1 NP_001329006.1 10 - 11 1508 61
NM_001341064.1 NP_001329005.1 10 - 11 1264 61
NM_001341065.1 NP_001329007.1 8 - 9 979 61
NM_117648.8 NP_193292.3 11 - 12 1301 61
GenBank mRNA EU915246.1 - 11 - 12 1301 61

View Full Product Details