Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAA3
Entrez Gene ID
827233
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT4G15570, DL3825W, FCAALL.334, MAGATAMA 3
Chromosome Location
Chr.4: 8892607 - 8899061 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341063.1 NP_001329006.1 19 - 20 2575 86
NM_001341064.1 NP_001329005.1 19 - 20 2331 86
NM_001341065.1 NP_001329007.1 17 - 18 2046 86
NM_117648.8 NP_193292.3 20 - 21 2368 86
GenBank mRNA EU915246.1 - 20 - 21 2368 86

View Full Product Details