Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAA3
Entrez Gene ID
827233
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT4G15570, DL3825W, FCAALL.334, MAGATAMA 3
Chromosome Location
Chr.4: 8892607 - 8899061 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341064.1 NP_001329005.1 2 - 3 329 102
NM_117648.8 NP_193292.3 3 - 4 366 102
GenBank mRNA EU915246.1 - 3 - 4 366 102

View Full Product Details