Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIP2;2
Entrez Gene ID
827446
Gene Name
tonoplast intrinsic protein 2;2
Gene Aliases
AT4G17340, DELTA-TIP2, DL4705W, FCAALL.412, tonoplast intrinsic protein 2;2
Chromosome Location
Chr.4: 9698940 - 9700472 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117838.4 NP_193465.1 1 - 2 755 61
GenBank mRNA AF367283.1 - 1 - 2 439 61
AY056063.1 - 1 - 2 377 61
AY088929.1 - 1 - 2 443 61

View Full Product Details