Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHYE
Entrez Gene ID
827538
Gene Name
phytochrome E
Gene Aliases
AT4G18130, F15J5.100, F15J5_100, PHYTOCHROME E, phytochrome E
Chromosome Location
Chr.4: 10042137 - 10046346 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117923.8 NP_193547.4 2 - 3 3202 95

View Full Product Details