Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RLP51
- Entrez Gene ID
-
827610
- Gene Name
- receptor like protein 51
- Gene Aliases
-
AT4G18760, AtRLP51, F28A21.170, F28A21_170, SNC2, constitutive 2, receptor like protein 51, suppressor of npr1-1
- Chromosome Location
-
Chr.4: 10307810 - 10309591 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117992.2 |
NP_193611.1 |
1 - 1 |
1258 |
142 |
View Full Product Details