The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_117994.4 | NP_567564.1 | 8 - 9 | 1524 | 128 |
GenBank mRNA | AY099636.1 | - | 8 - 9 | 1424 | 128 |
KJ676982.1 | - | 8 - 9 | 1357 | 128 |