Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FXG1
- Entrez Gene ID
-
843109
- Gene Name
- alpha-fucosidase 1
- Gene Aliases
-
AT1G67830, ATFXG1, Arabidopsis thaliana alpha-fucosidase 1, F12A21.4, F12A21_4, alpha-fucosidase 1
- Chromosome Location
-
Chr.1: 25431549 - 25433091 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105451.4 |
NP_176949.1 |
2 - 3 |
727 |
81 |
View Full Product Details