Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIS1
Entrez Gene ID
843128
Gene Name
phosphatidylinositol synthase 1
Gene Aliases
AT1G68000, ATPIS, ATPIS1, PHOSPHATIDYLINOSITOL SYNTHASE, T23K23.15, T23K23_15, phosphatidylinositol synthase 1
Chromosome Location
Chr.1: 25490911 - 25493077 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105470.3 NP_176967.1 4 - 5 504 76
GenBank mRNA AJ000539.1 - 4 - 5 341 76
AY140047.1 - 4 - 5 396 76

View Full Product Details