Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MINE1
Entrez Gene ID
843271
Gene Name
homologue of bacterial MinE 1
Gene Aliases
ARC12, AT1G69390, ATMINE1, F23O10.25, F23O10_25, accumulation and replication of chloroplasts 12, homologue of bacterial MinE 1
Chromosome Location
Chr.1: 26084562 - 26086512 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105606.4 NP_564964.1 1 - 2 262 83
GenBank mRNA AB046117.1 - 1 - 2 146 83
AY088828.1 - 1 - 2 170 83
BT002897.1 - 1 - 2 168 83
BT004467.1 - 1 - 2 103 83

View Full Product Details