Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EDA25
- Entrez Gene ID
-
843576
- Gene Name
- CCAAT-binding factor
- Gene Aliases
-
AT1G72440, SLOW WALKER2, SWA2, T10D10.9, T10D10_9, embryo sac development arrest 25
- Chromosome Location
-
Chr.1: 27268092 - 27273935 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105903.4 |
NP_177388.2 |
13 - 14 |
2186 |
135 |
View Full Product Details