Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTE3
Entrez Gene ID
843646
Gene Name
global transcription factor group E3
Gene Aliases
AT1G73150, F3N23.35, F3N23_35, global transcription factor group E3
Chromosome Location
Chr.1: 27503973 - 27506406 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105973.4 NP_177458.1 1 - 2 1609 131
GenBank mRNA BT020256.1 - 1 - 2 1231 131

View Full Product Details