Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BNS
Entrez Gene ID
843651
Gene Name
bonsai
Gene Aliases
APC13, AT1G73177, BONSAI, anaphase-promoting complex 13
Chromosome Location
Chr.1: 27516572 - 27517882 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105976.3 NP_565057.1 3 - 4 253 78
GenBank mRNA AY088589.1 - 3 - 4 225 78
BT025015.1 - 2 - 3 134 78

View Full Product Details