Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUS5
Entrez Gene ID
833692
Gene Name
sucrose synthase 5
Gene Aliases
ARABIDOPSIS THALIANA SUCROSE SYNTHASE 5, AT5G37180, ATSUS5, MJG14.14, MJG14_14, sucrose synthase 5
Chromosome Location
Chr.5: 14718141 - 14723139 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344166.1 NP_001318683.1 5 - 6 1089 107
NM_001344167.1 NP_001332704.1 1 - 2 494 107
NM_123077.3 NP_198534.3 3 - 4 669 107

View Full Product Details