Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRL5
Entrez Gene ID
821101
Gene Name
APR-like 5
Gene Aliases
APR-like 5, AT3G03860, ATAPRL5, F20H23.9, F20H23_9
Chromosome Location
Chr.3: 992074 - 994697 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111257.4 NP_566214.1 3 - 4 875 92
GenBank mRNA AY059721.1 - 3 - 4 751 92
AY091351.1 - 3 - 4 484 92

View Full Product Details