Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAR5
Entrez Gene ID
823580
Gene Name
fatty acid reductase 5
Gene Aliases
AT3G44550, fatty acid reductase 5
Chromosome Location
Chr.3: 16137576 - 16142270 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339154.1 NP_001325959.1 5 - 6 902 102
NM_114323.4 NP_190041.2 6 - 7 1327 102
GenBank mRNA AK228404.1 - 6 - 7 886 102

View Full Product Details