Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC13
Entrez Gene ID
815414
Gene Name
multidrug resistance-associated protein 11
Gene Aliases
AT2G07680, ATMRP11, ATP-binding cassette C13, AtABCC13, MRP11, multidrug resistance-associated protein 11
Chromosome Location
Chr.2: 3514668 - 3522658 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335346.1 NP_001323940.1 31 - 32 3598 115
NM_001335347.1 NP_001323936.1 30 - 31 3515 115
NM_001335348.1 NP_001323937.1 30 - 31 3444 115
NM_001335349.1 NP_001323938.1 28 - 29 3375 115
NM_126770.8 NP_178811.7 30 - 31 3455 115

View Full Product Details