Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GATL5
- Entrez Gene ID
-
839475
- Gene Name
- galacturonosyltransferase 5
- Gene Aliases
-
AT1G02720, AtGATL5, T14P4.1, T14P4_1, galacturonosyltransferase 5, galacturonosyltransferase-like 5
- Chromosome Location
-
Chr.1: 591682 - 594264 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_202015.2 |
NP_973744.1 |
1 - 1 |
1382 |
128 |
View Full Product Details