Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSZ21
Entrez Gene ID
838997
Gene Name
RS-containing zinc finger protein 21
Gene Aliases
AT1G23860, At-RSZ21, RS-containing zinc finger protein 21, RSZP21, SRZ-21, SRZ21, T23E23.2, T23E23_2
Chromosome Location
Chr.1: 8427257 - 8429498 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123870.4 NP_001117342.1 3 - 4 313 115
NM_001198145.1 NP_001185074.1 3 - 4 438 115
NM_102234.4 NP_564208.1 3 - 4 425 115
NM_202172.1 NP_973901.1 2 - 3 257 115
GenBank mRNA AF033587.1 - 3 - 4 238 115
AK317576.1 - 3 - 4 266 115

View Full Product Details