Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PR4
Entrez Gene ID
819632
Gene Name
pathogenesis-related 4
Gene Aliases
AT3G04720, AtPR4, F7O18.21, F7O18_21, HEL, HEVEIN-LIKE, PR-4, pathogenesis-related 4
Chromosome Location
Chr.3: 1285425 - 1286760 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111344.6 NP_187123.1 1 - 2 635 98
GenBank mRNA AF370536.1 - 1 - 2 432 98
AY088644.1 - 1 - 2 437 98
BT000046.1 - 1 - 2 406 98
U01880.1 - 1 - 2 411 98

View Full Product Details