Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LON4
Entrez Gene ID
819748
Gene Name
lon protease 4
Gene Aliases
AT3G05790, F10A16.8, F10A16_8, lon protease 4
Chromosome Location
Chr.3: 1719958 - 1725268 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337610.1 NP_001325548.1 10 - 11 1101 141
NM_111453.1 NP_566259.1 10 - 11 1048 141

View Full Product Details