Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPM2
Entrez Gene ID
819793
Gene Name
BTB-POZ and MATH domain 2
Gene Aliases
AT3G06190, ATBPM2, BTB-POZ AND MATH DOMAIN 2, BTB-POZ and MATH domain 2, F28L1.13, F28L1_13
Chromosome Location
Chr.3: 1874169 - 1876946 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337638.1 NP_001326823.1 3 - 4 1736 87
NM_001337639.1 NP_001326822.1 4 - 5 1462 87
NM_111494.3 NP_566275.1 3 - 4 1405 87
GenBank mRNA AY045891.1 - 3 - 4 1374 87
AY091449.1 - 3 - 4 1034 87

View Full Product Details