Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REP
Entrez Gene ID
819832
Gene Name
Rab escort protein
Gene Aliases
AT3G06540, AthREP, F5E6.13, F5E6_13, Rab escort protein
Chromosome Location
Chr.3: 2034913 - 2037791 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111530.4 NP_187306.2 7 - 8 1629 69
GenBank mRNA AF409086.2 - 7 - 8 1606 69

View Full Product Details