Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACLB-1
Entrez Gene ID
819845
Gene Name
ATP-citrate lyase B-1
Gene Aliases
AT3G06650, ATP-citrate lyase B-1, T8E24.7, T8E24_7
Chromosome Location
Chr.3: 2079033 - 2083472 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337675.1 NP_001326324.1 2 - 3 835 81
NM_111541.4 NP_187317.1 2 - 3 444 81
GenBank mRNA BT002736.1 - 1 - 2 101 81

View Full Product Details