Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ULP1A
- Entrez Gene ID
-
819876
- Gene Name
- UB-like protease 1A
- Gene Aliases
-
AT3G06910, AtULP1a, ELS1, ESD4-LIKE SUMO PROTEASE 1, UB-like protease 1A
- Chromosome Location
-
Chr.3: 2178630 - 2181341 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111571.3 |
NP_187347.2 |
6 - 7 |
1427 |
67 |
View Full Product Details