Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QRT2
Entrez Gene ID
819988
Gene Name
Pectin lyase-like superfamily protein
Gene Aliases
AT3G07970, QUARTET 2
Chromosome Location
Chr.3: 2540913 - 2543613 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111676.4 NP_187454.2 3 - 4 656 118

View Full Product Details