Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATR1
Entrez Gene ID
828554
Gene Name
P450 reductase 1
Gene Aliases
AR1, ARABIDOPSIS CYTOCHROME REDUCTASE, AT4G24520, F22K18.280, F22K18_280, P450 reductase 1
Chromosome Location
Chr.4: 12662561 - 12667262 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203894.1 NP_001190823.1 16 - 17 2117 61
NM_118585.4 NP_194183.1 16 - 17 2192 61
GenBank mRNA AY054688.1 - 16 - 17 2085 61
BT008426.1 - 16 - 17 1996 61

View Full Product Details