The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203894.1 | NP_001190823.1 | 5 - 6 | 703 | 92 |
NM_118585.4 | NP_194183.1 | 5 - 6 | 778 | 92 | |
GenBank mRNA | AY054688.1 | - | 5 - 6 | 671 | 92 |
BT008426.1 | - | 5 - 6 | 582 | 92 |