Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSB2
Entrez Gene ID
828815
Gene Name
tryptophan synthase beta-subunit 2
Gene Aliases
AT4G27070, T24A18.20, T24A18_20, TRYPTOPHAN SYNTHASE BETA-SUBUNIT, tryptophan synthase beta-subunit 2
Chromosome Location
Chr.4: 13586268 - 13588897 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118841.5 NP_194437.1 1 - 2 652 91
GenBank mRNA AK228129.1 - 1 - 2 408 91
AY084334.1 - 1 - 2 405 91
BT003144.1 - 1 - 2 356 91

View Full Product Details