Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAA11
Entrez Gene ID
828982
Gene Name
indole-3-acetic acid inducible 11
Gene Aliases
AT4G28640, T5F17.90, T5F17_90, indole-3-acetic acid inducible 11
Chromosome Location
Chr.4: 14142063 - 14143969 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084991.1 NP_001078460.1 2 - 3 687 123
NM_001203930.2 NP_001190859.1 2 - 3 759 123
NM_119006.3 NP_194593.1 2 - 3 687 123
GenBank mRNA AF332397.1 - 2 - 3 534 123
AK118377.1 - 2 - 3 673 123
U18413.1 - 2 - 3 577 123

View Full Product Details