Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPT2a
Entrez Gene ID
829025
Gene Name
regulatory particle AAA-ATPase 2A
Gene Aliases
26S PROTEASOME AAA-ATPASE SUBUNIT RPT2A, AT4G29040, F19B15.70, F19B15_70, regulatory particle AAA-ATPase 2A
Chromosome Location
Chr.4: 14312283 - 14314600 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119048.4 NP_194633.1 5 - 6 1337 103
GenBank mRNA AB161192.1 - 5 - 6 1317 103
AY034975.1 - 5 - 6 1317 103

View Full Product Details