Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD25
Entrez Gene ID
829036
Gene Name
IQ-domain 25
Gene Aliases
AT4G29150, F19B15.180, F19B15_180, IQ-domain 25
Chromosome Location
Chr.4: 14378706 - 14380512 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119059.4 NP_194644.2 1 - 2 494 85

View Full Product Details