Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATMND1
Entrez Gene ID
829038
Gene Name
Mnd1 family protein
Gene Aliases
AT4G29170, F19B15.200, F19B15_200
Chromosome Location
Chr.4: 14382819 - 14385465 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085000.2 NP_001078469.1 5 - 6 591 66
NM_001341959.1 NP_001329788.1 5 - 6 513 66
NM_119061.4 NP_194646.2 5 - 6 496 66
GenBank mRNA AK117713.1 - 5 - 6 445 66
AM162278.1 - 4 - 5 347 66
BT005435.1 - 4 - 5 347 66
DQ248000.1 - 5 - 6 468 66

View Full Product Details