Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG4
Entrez Gene ID
829210
Gene Name
SET domain group 4
Gene Aliases
ASH1-RELATED 3, ASH1-related 3, ASHR3, AT4G30860, F6I18.230, F6I18_230, SET domain group 4
Chromosome Location
Chr.4: 15024444 - 15027672 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119233.2 NP_567859.1 5 - 6 905 148
GenBank mRNA AY050894.1 - 5 - 6 877 148
AY096675.1 - 5 - 6 803 148

View Full Product Details