Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EXO70G1
- Entrez Gene ID
-
829281
- Gene Name
- exocyst subunit exo70 family protein G1
- Gene Aliases
-
AT4G31540, ATEXO70G1, F3L17.110, F3L17_110, exocyst subunit exo70 family protein G1
- Chromosome Location
-
Chr.4: 15284385 - 15287464 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119303.3 |
NP_194882.2 |
1 - 1 |
2908 |
115 |
- Gene Symbol
- AT4G31530
- Entrez Gene ID
-
829280
- Gene Name
- NAD(P)-binding Rossmann-fold superfamily protein
- Gene Aliases
-
AT4G31530, F3L17.100, F3L17_100
- Chromosome Location
-
Chr.4: 15282230 - 15287390 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119302.5 |
NP_194881.2 |
9 - 9 |
1519 |
115 |
View Full Product Details