Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATM2
Entrez Gene ID
835516
Gene Name
myosin 2
Gene Aliases
ARABIDOPSIS THALIANA MYOSIN 1, ARABIDOPSIS THALIANA MYOSIN 4, AT5G54280, ATM4, ATMYOS1, MDK4.10, MDK4_10, MYOSIN 4, myosin 2
Chromosome Location
Chr.5: 22039383 - 22046358 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085286.2 NP_001078755.1 18 - 19 2792 114
NM_124808.4 NP_568806.3 17 - 18 1916 114

View Full Product Details