Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT10
Entrez Gene ID
828648
Gene Name
nudix hydrolase homolog 10
Gene Aliases
AT4G25434, ATNUDT10, nudix hydrolase homolog 10
Chromosome Location
Chr.4: 13003568 - 13005935 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036636.2 NP_001031713.1 3 - 4 539 67
NM_001341737.1 NP_001329743.1 1 - 2 893 67
NM_001341742.1 NP_001329747.1 3 - 4 503 67
NM_179112.3 NP_849443.2 3 - 4 503 67
GenBank mRNA BT010765.1 - 3 - 4 467 67
BT010987.1 - 3 - 4 375 67

View Full Product Details