Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATU2AF65A
Entrez Gene ID
829822
Gene Name
U2 snRNP auxilliary factor, large subunit, splicing factor
Gene Aliases
AP22.29, AP22_29, AT4G36690
Chromosome Location
Chr.4: 17293938 - 17297761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204008.1 NP_001190937.1 12 - 13 1667 75
NM_119833.4 NP_195387.1 12 - 13 1700 75
NM_179178.3 NP_849509.1 11 - 12 1967 75
NM_202966.4 NP_974695.1 12 - 13 1867 75
GenBank mRNA AF462805.1 - 12 - 13 1694 75
AY080711.1 - 11 - 12 1966 75
AY143980.1 - 12 - 13 1581 75
BT000965.1 - 12 - 13 1581 75

View Full Product Details