Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATU2AF65A
Entrez Gene ID
829822
Gene Name
U2 snRNP auxilliary factor, large subunit, splicing factor
Gene Aliases
AP22.29, AP22_29, AT4G36690
Chromosome Location
Chr.4: 17293938 - 17297761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204008.1 NP_001190937.1 7 - 8 841 78
NM_119833.4 NP_195387.1 7 - 8 874 78
NM_179178.3 NP_849509.1 7 - 8 868 78
NM_202966.4 NP_974695.1 7 - 8 868 78
GenBank mRNA AF462805.1 - 7 - 8 868 78
AY080711.1 - 7 - 8 867 78
AY143980.1 - 7 - 8 755 78
BT000965.1 - 7 - 8 755 78

View Full Product Details