Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CGL1
Entrez Gene ID
829981
Gene Name
alpha-1,3-mannosyl-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Gene Aliases
AT4G38240, CGL, COMPLEX GLYCAN LESS, COMPLEX GLYCAN LESS 1, F20D10.360, F20D10_360, GNTI, N-ACETYLGLUCOSAMINYLTRANSFERASE I
Chromosome Location
Chr.4: 17931570 - 17935325 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204022.1 NP_001190951.1 7 - 8 519 72
NM_119986.3 NP_195537.2 7 - 8 543 72
NM_179186.1 NP_849517.1 7 - 8 543 72
GenBank mRNA AJ243198.1 - 7 - 8 457 72
AY099838.1 - 7 - 8 514 72
BT000334.1 - 7 - 8 427 72
KJ138715.1 - 7 - 8 427 72

View Full Product Details