Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCCA
Entrez Gene ID
838362
Gene Name
methylcrotonyl-CoA carboxylase alpha chain
Gene Aliases
AT1G03090
Chromosome Location
Chr.1: 739544 - 744184 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179252.3 NP_849583.1 6 - 7 1069 63
GenBank mRNA AY070723.1 - 6 - 7 926 63

View Full Product Details