Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCCA
Entrez Gene ID
838362
Gene Name
methylcrotonyl-CoA carboxylase alpha chain
Gene Aliases
AT1G03090
Chromosome Location
Chr.1: 739544 - 744184 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100191.4 NP_563674.1 7 - 8 1078 104
NM_179252.3 NP_849583.1 8 - 9 1272 104
GenBank mRNA AY070723.1 - 8 - 9 1129 104

View Full Product Details