Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UVH6
Entrez Gene ID
839557
Gene Name
RAD3-like DNA-binding helicase protein
Gene Aliases
ARABIDOPSIS THALIANA XERODERMA PIGMENTOSUM GROUP D, AT1G03190, ATXPD, ULTRAVIOLET HYPERSENSITIVE 6
Chromosome Location
Chr.1: 775527 - 780062 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100201.3 NP_171818.1 3 - 4 953 91
NM_179253.1 NP_849584.1 2 - 3 1016 91
GenBank mRNA AY090788.1 - 2 - 3 1016 91
AY090915.1 - 2 - 3 816 91

View Full Product Details