Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HUA1
Entrez Gene ID
820448
Gene Name
floral homeotic protein (HUA1)
Gene Aliases
AT3G12680, ENHANCER OF AG-4 1, T2E22.1
Chromosome Location
Chr.3: 4025076 - 4029140 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112103.3 NP_187874.2 5 - 6 962 122
GenBank mRNA AY024357.1 - 5 - 6 962 122

View Full Product Details