Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDRB
Entrez Gene ID
820462
Gene Name
short-chain dehydrogenase-reductase B
Gene Aliases
AT3G12790, AT3G12800, DECR, short-chain dehydrogenase-reductase B
Chromosome Location
Chr.3: 4063320 - 4064859 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112116.4 NP_187886.2 1 - 2 288 63
GenBank mRNA AK175259.1 - 1 - 2 217 63
AK176181.1 - 1 - 2 217 63
AK176305.1 - 1 - 2 214 63
AK176508.1 - 1 - 2 214 63
AK176776.1 - 1 - 2 194 63
AK176892.1 - 1 - 2 198 63
AK176909.1 - 1 - 2 214 63
AK221326.1 - 1 - 2 194 63
AK229784.1 - - 217 63
AY072396.1 - 1 - 2 216 63
AY114700.1 - 1 - 2 186 63

View Full Product Details