Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SRF4
- Entrez Gene ID
-
820494
- Gene Name
- STRUBBELIG-receptor family 4
- Gene Aliases
-
AT3G13065, MGH6.19, STRUBBELIG-receptor family 4
- Chromosome Location
-
Chr.3: 4187371 - 4191298 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112145.4 |
NP_566444.4 |
9 - 10 |
854 |
117 |
GenBank mRNA |
AY518289.1 |
- |
9 - 10 |
854 |
117 |
- Gene Symbol
- AT3G13062
- Entrez Gene ID
-
820493
- Gene Name
- Polyketide cyclase/dehydrase and lipid transport superfamily protein
- Gene Aliases
-
AT3G13062
- Chromosome Location
-
Chr.3: 4184340 - 4187453 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FJ154118.1 |
- |
15 - 16 |
2389 |
117 |
View Full Product Details