Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WIP3
Entrez Gene ID
820536
Gene Name
WPP domain interacting protein 3
Gene Aliases
AT3G13360, WPP domain interacting protein 3
Chromosome Location
Chr.3: 4338250 - 4340446 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338045.1 NP_001327578.1 2 - 3 1053 141
NM_112181.5 NP_566455.1 2 - 3 1037 141
GenBank mRNA AY045697.1 - 2 - 3 986 141
AY149963.1 - 1 - 2 866 141

View Full Product Details